Z675474
Sterile economy sample spoon
8 mL
Synonym(e):
dispensing tool
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122403
NACRES:
NB.35
Empfohlene Produkte
Beschreibung
PS
Hersteller/Markenname
Bel-Art F36740-00004
Verwandte Kategorien
Allgemeine Beschreibung
Moulded of lightweight and rigid, white high impact polystyrene. Individually packaged sterile spoons for one time use packed 100 per box.
- Use to collect specific volumes of dry or liquid materials
- Remains upright when filled or empty for use as a weighing boat
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z675474-100EA | 4061832266541 |