Merck
Z657220
IKA® overhead stirrer accessories
R271 boss head clamp
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.72
Verpackung
pack of 1 ea
Hersteller/Markenname
IKA 0002664000
Allgemeine Beschreibung
The boss head is used for the safe fixing of drives to the stands. It resists strongest mechanical demands.
Stand diam. 34 mm, Bracket diam. 16mm
Rechtliche Hinweise
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)