Z403628
IKA® disperser clamps
RH 3 Strap clamp for dispersers, 1/cs
Synonym(e):
homogenizer clamps, tissue grinder clamps
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.76
Empfohlene Produkte
Verpackung
pack of 1
Hersteller/Markenname
IKA 0003008600
Allgemeine Beschreibung
For securing vessels against walls or for synchronizing rotation during stirring or dispersing
- For stand diameter: 9-16mm
- For vessel diameter: 40-300mm
Rechtliche Hinweise
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)