Z569828
2mag MIXcontrol 20 control unit for MIXdrive
AC/DC input 100 - 240 V AC, Schuko plug
Synonym(e):
control unit, power bank, power unit
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103806
NACRES:
NB.72
Empfohlene Produkte
Leistungsmerkmale
CE compliant
Verpackung
pack of 1 ea
Hersteller/Markenname
2mag 90200
Leistungsmerkmale und Vorteile
- Control unit with 20 watts power
- Speed range from 100 up to 1,600 rpm
- Jerk-free stirring also at low speeds
- Power setting (4-steps) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation of MIXdrive stirring drives
- Clear digital display for speed- and power settings
- SoftStart for reliable catching/centering and safe accelerating of the stirring bar
- QuickSet for quick setting of start- and maximum speed, automatic memory function to save the last settings, resistant stainless steel housing, easy to clean
Zubehör
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z569828-1EA | 4061833479162 |