Z567604
Aldrich® swivel Benchclamp™
dual adjustment grippy knobs, 3-prong, size L
Synonym(e):
bench clamps
Alle Fotos(3)
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.35
Empfohlene Produkte
Größe
L
Backenkapazität
0-60 mm
Zinke B
8 mm
Allgemeine Beschreibung
Aldrich benchclamp clamps have a nickel-plated finish and are corrosion resistant. The clamps come with slip-on fiberglass finger covers. The Grippy Knob design has a round nut which is easy to turn making tightening the clamps around glassware fast and convenient. This swivel head allows for the clamp to have any angle of adjustment.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Benchclamp is a trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z567604-1EA | 4061833305782 |