Z256714
Kontes® threaded MICROFLEX® kits
Deluxe Academic kit
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
Empfohlene Produkte
Hersteller/Markenname
Kimble®
Allgemeine Beschreibung
Components Kit qty
A Claisen adapter, 13-425 1
B Connecting adapter, 13-425 1
C Air condenser, 13-425, 100mm 1
D Connector, 13-425 to 13-425 4
E Connector, 13-425 to 20-400 2
F Water-jacketed condenser, 13-425 1
G Craig recrystallization tube, 2 to 3mL 1
H Craig recrystallization plug, glass 1
I Hickman distillation head 1
J Drying tube 1
K Pear-shaped flask, 5mL 1
L Cylindrical flask, 10mL 1
Components Kit qty
M Micro vial, 0.1mL 1
N Micro vial, 3mL 1
O Micro vial, 5mL 1
P GC collection tube, 1
with size 006 Viton O-ring
Q Compression cap, 13-425 1
R Cap O-ring, Viton®, size 107 2
S Syringe, PE 1
T Gas delivery tube, 13-425 1
U Centrifuge tube, 15mL, 1
Foam insert set, 1
Ekono-Case 1
A Claisen adapter, 13-425 1
B Connecting adapter, 13-425 1
C Air condenser, 13-425, 100mm 1
D Connector, 13-425 to 13-425 4
E Connector, 13-425 to 20-400 2
F Water-jacketed condenser, 13-425 1
G Craig recrystallization tube, 2 to 3mL 1
H Craig recrystallization plug, glass 1
I Hickman distillation head 1
J Drying tube 1
K Pear-shaped flask, 5mL 1
L Cylindrical flask, 10mL 1
Components Kit qty
M Micro vial, 0.1mL 1
N Micro vial, 3mL 1
O Micro vial, 5mL 1
P GC collection tube, 1
with size 006 Viton O-ring
Q Compression cap, 13-425 1
R Cap O-ring, Viton®, size 107 2
S Syringe, PE 1
T Gas delivery tube, 13-425 1
U Centrifuge tube, 15mL, 1
Foam insert set, 1
Ekono-Case 1
Rechtliche Hinweise
KIMBLE is a registered trademark of DWK Life Sciences
Kontes is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
MICROFLEX is a registered trademark of Kimble Kontes Asset Mgmt. Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)