Merck
WHA10477602
Whatman® Vacuum filtration apparatus accessories
PZ 001 tweezers, stainless steel
Synonym(e):
Z751979
About This Item
UNSPSC-Code:
41104900
NACRES:
NB.24
Verpackung
pk of 1
Hersteller/Markenname
Whatman 10477602
Whatman Article No. 28415991 (US reference)
Allgemeine Beschreibung
The stainless-steel forceps with smooth angled jaws (104 mm long) are excellent for handling membrane filters.
Leistungsmerkmale und Vorteile
  • They are autoclavable and can be flame sterilized with ethanol.
Sonstige Hinweise
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Rechtliche Hinweise
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
WHA104776024061826076903