Merck
WHA10418712
Whatman® Nuclepore Track-Etched Membranes
polycarbonate filter, filter diam. 47 mm, pore size 1 μm, non-sterile, case of 100 ea
About This Item
UNSPSC-Code:
41104921
NACRES:
NB.24
Materialien
polycarbonate filter
Sterilität
non-sterile
Leistungsmerkmale
hydrophilic
Verpackung
case of 100 ea
Hersteller/Markenname
Cytiva 10418712
Durchmesser
47 mm
Filterdurchm.
47 mm
Porengröße
1 μm
Anwendung
  • Epifluorescence microscopy
  • Environmental analysis
  • Cell biologyEPA testing
  • Fuel testing
  • Bioassays
  • Parasitology
  • Air analysis
  • Water microbiology
Rechtliche Hinweise
Nuclepore is a trademark of Cytiva
Whatman is a registered trademark of Cytiva
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)