TEM-FF200CUUL
Ultrathin Formvar Supported Copper Square Mesh
size 200 mesh, box of 50
Synonym(e):
FORMVAR SQUARE MESH - CU 200 MESH UL 50/BX
Alle Fotos(1)
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Empfohlene Produkte
Verpackung
box of 50
Größe
200 mesh
Verwandte Kategorien
Allgemeine Beschreibung
A 5-6 nm ultrathin formvar film supported standard copper square mesh grids. Copper grid, 200 mesh. All of our film is laid on the shiny side of the grid.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)