Merck
TEM-CF200HCU
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 200 mesh, box of 25
Synonym(e):
CARBON FILM ONLY ON 200 HEX COPPER GRIDS 25/BX
About This Item
UNSPSC-Code:
41111761
NACRES:
MA.24
Materialien
carbon film
copper grid
Beschreibung
film layer thickness 5 - 6 nm carbon
Verpackung
box of 25
Größe
200 mesh
Allgemeine Beschreibung
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 200 mesh. Carbon film show better electrical and heat conduction in the electron beam.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)