Merck
SYNF671425
Synthware pressure equalizing addition funnel with PTFE stopcock in equalizing line
25 mL, joint: ST/NS 14/20, stopcock: PTFE
Synonym(e):
Synthware F671425, addition funnel
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.51
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F671425
Kapazität
25 mL
25 mL
Allgemeine Beschreibung
With straight bore PTFE stopcock in pressure equalizing line, graduated.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)