Merck
SYNF601002C
Synthware Buchner funnel
2 mL, porosity: coarse
Synonym(e):
Synthware F601002C, filtering funnel
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.51
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F601002C
Kapazität
2 mL
2 mL
Allgemeine Beschreibung
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)