Merck
SYNF549250
Synthware reaction flask with high vacuum valve
250 mL, joint: ST/NS 19/22, valve size 0-4 mm
Synonym(e):
Synthware F549250, Schlenk flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
borosilicate glass flask
round bottom
single-neck (high vaccum valve)
Verbindung
joint: ST/NS19/22
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware F549250
Kapazität
250 mL
250 mL
Ventilgröße
0-4 mm
Allgemeine Beschreibung
Supplied with high vacuum valve with PTFE O-ring at tip and PTFE wiper to protect Viton® O-ring on shaft.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)