SYNA391420
Synthware™ transfer valve adapter
joint: ST/NS 14/20
Synonym(e):
Synthware A391420, transfer adapter with valve
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.51
Empfohlene Produkte
Verbindung
joint: ST/NS14/20
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware A391420
Allgemeine Beschreibung
The 0-4 mm high vacuum valves have two Viton® O-rings on the shaft and an integral PTFE O-ring on the tip.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)