Merck
SLW2180/02M
Pyrex® Separating funnel, pear-shape, glass key
50 mL, glass stopper, PE stopper, pack of 1 ea
Synonym(e):
Pyrex, SciLabware 2180/02M
About This Item
UNSPSC-Code:
41121809
NACRES:
NB.43
Materialien
PE stopper
PTFE key
glass stopper
pear-shaped
Verbindung
joint ST/NS cone19/26
Verpackung
pack of 1 ea
Hersteller/Markenname
SciLabware 2180/02M
Trichterdurchm.
53 mm
Absperrhahnbohrung
2.0 mm
Kapazität
50 mL
Allgemeine Beschreibung
In liquid-liquid extractions, the two immiscible liquids separate into different layers where the liquid with the higher density collects in the lower part of the vessel and can be drained off via the stopcock.
Leistungsmerkmale und Vorteile
  • Complies with ISO 4800 Type 2
  • Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
  • With interchangeable PTFE stopcock key
  • Supplied with both glass and polyethylene stoppers
  • The 5Litre size is supplementary to the ISO range.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)