Merck
PL800GA-BUT-7
Plas-Labs replacement butyl gloves
for 2 piece glove set, size 7 (for 8 in. ports)
About This Item
UNSPSC-Code:
46181504
NACRES:
NB.35
Materialien
butyl rubber
Größe
7 (for 8 in. ports)
Leistungsmerkmale
reusable
for use with glove box sleeves
Verpackung
pack of 1 pair
Hersteller/Markenname
Plas-Labs 800GA-BUT-7
Allgemeine Beschreibung
Plas-Labs black butyl rubber gloves for highest impermeability to water vapor, gases, and toxic fluids
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
PL800GA-BUT-7-1PR4061832875392