MT59904591
Mettler Toledo RedCap pH electrode easy clean
L 120 mm × 12 mm
Alle Fotos(1)
About This Item
UNSPSC-Code:
41115604
NACRES:
NB.79
Empfohlene Produkte
Materialien
glass (A41)
glass membrane (polymer)
Beschreibung
S7 connection
Without cable
Hersteller/Markenname
Mettler-Toledo 59904591
L × B
120 mm × 12 mm
Länge
120 mm × 12 mm
Allgemeine Beschreibung
Mettler Toledo RedCap pH electrode 405-60-T-S7/120/9848 is a generalist. It can be used as a puncture electrode in soft media, but also for general aqueous samples. Thanks to the triple ceramic junction and huge glass membrane the 405-60-T-S7/120/9848 responds really quickly. The electrolyte is free from silver due to the ARGENTHAL™ reference system and the silver ion trap.
- Combination pH electrode
- Easy to clean cone-shaped tip
- Triple ceramic diaphragm
- Large A41 glass membrane
- Detachable cable
Rechtliche Hinweise
ARGENTHAL is a trademark of Mettler-Toledo, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
MT59904591-1EA |