Merck
DWK90K16125F
KIMBLE® disposable screw thread culture tube
flat bottom borosilicate glass, tube diam. × L (16 mm) (125 mm), non-sterile
Synonym(e):
90K16125F
About This Item
UNSPSC-Code:
41121706
NACRES:
NB.11
Materialien
flat bottom borosilicate glass
flat bottom
screw top neck
transparent borosilicate glass tube
Sterilität
non-sterile
Leistungsmerkmale
without cap
Verpackung
case of 1000 ea Modular tray shrink-wrapped in plastic film
Hersteller/Markenname
Kimble® 90K16125F
Röhrchendurchm. × L
(16 mm) (125 mm)
Fitting
neck (External Thread)
Allgemeine Beschreibung
Disposable Screw Thread Culture Tubes
  • These tubes are suitable for tissue culture work and general bacteriological use.
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
  • No marking spot
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Leistungsmerkmale und Vorteile
  • Manufactured from borosilicate glass 5.1
  • 73760 series and 90 series items that end in ‘F′ have flat bottoms which contribute to stability
  • 73770 series items are produced with a round bottom
  • Modular trays are shrink-wrapped in plastic film
  • Supplied without caps
  • No marking spot
Rechtliche Hinweise
KIMBLE is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)