Merck
CLS6159613L
PYREXPLUS® coated solution bottles
capacity 13.25 L
Synonym(e):
Corning® 61596
About This Item
UNSPSC-Code:
41121800
Kapazität:
13.25 L
Hersteller/Markenname
Corning 61596-13L
Kapazität
13.25 L
Allgemeine Beschreibung
Features a protective plastic coating that helps prevent glass from shattering and reduces any spills. Fully autoclavable (121°C) and resistant to thermal shock. Graduated in 500mL increments with black enamel. Uses rubber stopper no. 12.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREXPLUS is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)