Merck
CLS560050
Pyrex® Lifetime-Red volumetric flask with snap cap
capacity 50 mL
Synonym(e):
clear flasks, pyrex flask, volumetric flasks, Corning® 5600
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
long
Sterilität
non-sterile
Leistungsmerkmale
closure type Snap cap
Verpackung
case of 12
pack of 6
Hersteller/Markenname
Corning 5600-50
tol. measuring range
0.05 mL
Kapazität
50 mL
Allgemeine Beschreibung
Capacity mark is fine line-etched through narrow, red band into clear glass.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Lifetime-Red is a trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)