Merck
BR92773
BRAND® Erlenmeyer flask with beaded rim and graduation, narrow mouth
volume 5000 mL
Synonym(e):
Erlenmeyer flasks, Flasks
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
flat bottom
Verbindung
joint neck52 mm
Sterilität
non-sterile
Leistungsmerkmale
without cap
Verpackung
pack of 10 ea
Hersteller/Markenname
BRAND 92773
Kapazität
5000 mL
Volumen
5000 mL
Allgemeine Beschreibung
The BRAND® Erlenmeyer Flask with beaded rim and graduations is a high-quality laboratory vessel designed for versatile use in chemical and biological applications. Made from Boro 3.3 glass, this flask meets DIN ISO 1773 standards, ensuring durability and reliability in various laboratory environments.
Leistungsmerkmale und Vorteile
  • Good mechanical resistance
  • Easy to clean
  • Diverse applications
Sonstige Hinweise
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)