Merck
BR87826
BRAND® PP beaker with spout, low form
volume 5000 mL
Synonym(e):
Beakers, Plastics
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
Materialien
polypropylene
Beschreibung
ISO 7056
Leistungsmerkmale
graduations
beaded rim and spout
Hersteller/Markenname
BRAND 87826
Volumen
5000 mL
Allgemeine Beschreibung
The BRAND® PP Beaker with Spout in low form is crafted from high-clarity polypropylene (PP) and complies with ISO 7056 standards, ensuring durability and reliability for laboratory applications. This beaker features clear graduation markings for precise measurement, along with a beaded rim that enhances strength and provides a comfortable grip. The integrated spout allows for easy pouring, minimizing spills and improving accuracy during liquid transfer. To preserve the marks and inscriptions, it is recommended to clean the beaker at temperatures up to a maximum of 60 °C.
Sonstige Hinweise
Browse all plastic beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
BR87826-4EA4061831848021