Merck
BR718020
BRAND® counting chamber BLAUBRAND® Thoma pattern
with clips, double ruled
Synonym(e):
BLAUBRAND®, Cell culture, Counting Chambers, Thoma
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
Materialien
colorless
glass
Sterilität
non-sterile
Leistungsmerkmale
with clips
Hersteller/Markenname
BRAND 718020
Wells
1
Allgemeine Beschreibung
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.Rulings correspond to the central large square of the Neubauer chamber. The mini squares have an area of 0.0025 mm² each. Since the outer large squares are not completed, the Thoma chamber is only used for counting thrombocytes and erythrocytes.
Leistungsmerkmale und Vorteile
• BLAUBRAND® quality• Outstanding measurement precision• 100% certified quality
Verpackung
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Rechtliche Hinweise
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)