Merck
BR701967
BRAND® repair set for TRANSFERPETTOR
for 50 μL
Synonym(e):
Accessories, Pipettes, Piston-operated pipettes, Positive displacement principle, Repair set, Transferpettor
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.01
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 701967
Verwandte Kategorien
Allgemeine Beschreibung
Includes: 1 allen key, 1 piston rod with fitted PTFE Transferpettor-seal (above size 20 μL), 1 calibrating gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 Transferpettor-seals, PTFE 1 mounting block (above size 20 μL).
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)