Z678147
Finnpipette® F3 variable volume pipette
volume 100-1000 μL
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Empfohlene Produkte
Leistungsmerkmale
color-coded blue
electronic: no
single-channel
Verpackung
pack of 1 ea
Hersteller/Markenname
Finn 4640060
Volumen
100-1000 μL
Verwandte Kategorien
Allgemeine Beschreibung
Finnpipette F3 has a double action pipetting button with rotating upper part enabling smooth and steady pipetting action at the same time as preventing accidental volume alterations. Finpipette F3 is equipped with the Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
- Double action pipetting button with very light and smooth pipetting action
- Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
- Big display with white numbers on black background for increased visibility
- Color coded for easy tip volume recognition
Rechtliche Hinweise
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z678147-1EA | 4061833036068 |