Merck
BR137935
BRAND® magnetic stirring bar, PTFE, double-ended
double-ended, L 54 mm, diam. 8 mm
Synonym(e):
Magnetic Stirring Bars, PTFE
About This Item
UNSPSC-Code:
41122401
NACRES:
NB.72
Materialien
PTFE
double-ended
Hersteller/Markenname
BRAND 137935
Länge
54 mm
Durchmesser
8 mm
Allgemeine Beschreibung
Excellent centering, small contact surface and high turbulence even at low speeds.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)