Merck
BR114105
BRAND® culture tube, rimless
L × W 10 mm × 75 mm, thickness 0.6 mm
Synonym(e):
Culture tubes, Tubes
About This Item
UNSPSC-Code:
41121706
NACRES:
NB.22
Materialien
round bottom
screw top cap
soda-lime glass tube
straight neck
transparent tube
Sterilität
non-sterile
Leistungsmerkmale
RCF 3,000 × g
neck thread push fit
Verpackung
pack of 250 ea
Hersteller/Markenname
BRAND 114105
L × B
10 mm × 75 mm
Kapazität
10 mL
3 mL
Dicke
0.6 mm
Allgemeine Beschreibung
Crafted from high-quality glass, these tubes provide excellent clarity and durability, making them ideal for cell culture and other scientific applications. Autoclavable at 121 °C, ensuring easy sterilization.
Leistungsmerkmale und Vorteile
  • Excellent sample visibility
  • Good chemical resistance
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)