Skip to Content
Merck
All Photos(1)

Key Documents

CF0004

Sigma-Aldrich

Alcohol Togni-(PhSCF2CF2)-reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H15F4IOS
CAS Number:
MDL number:
UNSPSC Code:
12352200

form

solid

reaction suitability

reaction type: C-C Bond Formation

storage temp.

−20°C

Application

Alcohol Togni-(PhSCF2CF2)-reagent is an electrophilic hypervalent iodine-fluoroalkyl reagent introducing the phenylsulfanyltetrafluoroethyl fragment. A phenylsulfanyltetrafluoroethyl moiety incorporated in the substrate can be treated with tributyltin hydride and generate the corresponding fluoroalkyl radical. If the substrate lacks any olefine, it can be reduced to tetrafluoroethyl group whereas if the substrate contains an olefine in the correct spatial orientation, it can lead to an intramolecular cyclisation affording tetrafluorinated cyclic structures.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The fluoroalkylation toolbox now includes Togni reagents, hypervalent iodine perfluoroalkylation reagents, fluoroalkyl bromides, silanes, carboxylates, and sulfonyl fluorides for late stage fluoroalkylation.

The fluoroalkylation toolbox now includes Togni reagents, hypervalent iodine perfluoroalkylation reagents, fluoroalkyl bromides, silanes, carboxylates, and sulfonyl fluorides for late stage fluoroalkylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service