Cultivo celular primario
Descripción general de las células primarias
Las células primarias son células de mamífero que se recolectan directamente de los tejidos vivos humanos o de animales. Difieren de los citoblastos en que las células primarias no citoblásticas están terminalmente diferenciadas y, por tanto, se consideran muy representativas de los tejidos de los que se han aislado, en particular cuando las características de la biología del cáncer pueden hacer que las líneas celulares cancerosas constituyan modelos menos predictivos de los sistemas normales o de las enfermedades no cancerosas. Los fenotipos comunes de las células primarias son: epiteliales, endoteliales, queratinocitos, melanocitos, fibroblastos, osteoblastos, miocitos y hepatocitos.
Artículos técnicos relacionados
- What are primary cells? A comprehensive overview of animal and human primary cell lines and media and how they are used in biomedical cell culture applications. Primary cultures are less homogeneous relative to cell lines.
- Primary human fibroblasts and growth medium for the in vitro cultivation of juvenile and adult fibroblasts. Protocols for handling, cell passaging, media, and product use.
- The PromoCell Primary Cancer Culture System allows selective isolation of malignant cancer cells from tumor associated stromal cells in patient derived xenographs (PDX models) and tumor samples.
- Enzyme Explorer Key Resource: Collagenase Guide.Collagenases, enzymes that break down the native collagen that holds animal tissues together, are made by a variety of microorganisms and by many different animal cells.
- Primary human airway epithelial cells and serum-free growth media are optimized for the in vitro cultivation of epithelial cells from large air passages. Protocols for handling, cell passaging, media, and product use.
- Ver todo (35)
Protocolos relacionados
- Human Umbilical Vein Endothelial Cells (HUVEC) Culture Protocol. Storage. Preparation for Culturing. Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.
- Ficoll 400® is a highly branched polymer formed by the copolymerization of sucrose and epichlorohydrin. Ficoll 400® is completely non-ionic. Because of the abundance of hydroxyl groups, Ficoll 400® is very hydrophilic and extremely water-soluble.
- Generate phagocytic M1 and M2 macrophages from human PBMCs in 10 days in serum-free and xeno-free cell culture media. Explore over 350 PromoCell products.
- Osteogenesis, adipogenesis and chondrogenesis differentiation protocols and media for human mesenchymal stem cell culture. Explore over 350 PromoCell products on our company website.
- Store the cryovials in a liquid nitrogen storage tank immediately upon arrival.
- Ver todo (62)
Encuentre más artículos y protocolos
Cómo difieren las células primarias de las líneas celulares inmortalizadas
Igual que las líneas celulares derivadas de tumores o inmortalizadas por otros métodos, las células primarias de origen humano o animal proporcionan un método de modelado de los sistemas biológicos. Debido a que suelen ser viables durante menos duplicaciones, las células primarias son menos sensibles a la pérdida de rasgos histoespecíficos que puede caracterizar a las células cultivadas durante períodos prolongados y pases repetidos in vitro. Las células primarias pueden documentarse con las características de los donantes, como edad, género y salud, cuando proceda, y por consiguiente pueden resultar modelos útiles para el desarrollo de enfoques de medicina personalizada.
Obtención y cultivo de células primarias
Las instituciones de investigación asociadas a hospitales pueden tener acceso a células y tejidos primarios. Los proveedores de células primarias ofrecen lotes celulares procedentes de donantes individuales o células combinadas de varios donantes. Los proveedores acreditados o los protocolos de recogida de pacientes deben incluir documentación que demuestre la supervisión ética del aislamiento de las células de los donantes humanos, así como la validación del tipo de célula. Las células primarias difieren de las líneas celulares de mamífero en que son generalmente más exigentes y requieren nutrientes añadidos, en particular cuando se cultivan en medios que tienen poco suero o ninguno. Estos nutrientes añadidos definidos son las citocinas y los factores de crecimiento.
Para seguir leyendo, inicie sesión o cree una cuenta.
¿No tiene una cuenta?