Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300550

Sigma-Aldrich

Anti-IKBKB (Ab-199) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FLJ40509 antibody produced in rabbit, Anti-IKK-beta antibody produced in rabbit, Anti-IKK2 antibody produced in rabbit, Anti-IKKB antibody produced in rabbit, Anti-inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~87 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200

isotype

IgG

immunogen sequence

(Q-K-Y-T-V)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IKBKB(3551)

Immunogen

Peptide sequence around aa. 197-201 (Q-K-Y-T-V), according to the protein IKBKB.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Acts as part of the IKK complex in the conventional pathway of NF-kappa-B activation and phosphorylates inhibitors of NF-kappa-B thus leading to the dissociation of the inhibitor/NF-kappa-B complex and ultimately the degradation of the inhibitor. Also phosphorylates NCOA3.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service