Skip to Content
Merck
All Photos(1)

Key Documents

MAB4301X

Sigma-Aldrich

Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480, Alexa Fluor 488

clone MC-480, Chemicon®, from mouse

Synonym(s):

SSEA-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MC-480, monoclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable

input

sample type: human embryonic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type neural stem cell(s)

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FUT4(2526)

General description

SSEA-1 is expressed on the surface of early mouse embryos, murine embryonal carcinoma cells (EC), murine embryonic stem cells (ES) and murine & human germ cells (EG). No immunoreactivity is evident with undifferentiated human EC and ES cells. Expression of SSEA-1 is down regulated following differentiation of murine EC and ES cells. In contrast, the differentiation of human EC and ES cells is characterized by an increase in SSEA-1 expression.

Specificity

Reacts with the Stage-specific embryonic antigen-1 (SSEA-1) that is expressed upon the surface of early mouse embryos, murine embryonal carcinoma cells (EC), murine embryonic stem cells (ES) and murine & human germ cells (EG). No immunoreactivity is evident with undifferentiated human EC and ES cells. Expression of SSEA-1 is down regulated following differentiation of murine EC and ES cells. In contrast, the differentiation of human EC and ES cells is characterized by an increase in SSEA-1 expression.

Immunogen

F9 teratocarcinoma stem cells (X-irradiated)

Application

FACS

Immunocytochemistry:

1. Fix cells with a 4% Paraformaldehyde solution for 30-45 minutes, and then rinse several times with Tris-Buffered-Saline (TBS).

2. Incubate cells with MAB4301X diluted to 1/100 with PBS/ Tween-20 (0.05%) / BSA (1%) for 30 minutes.

3. Wash the sample three times with PBS / Tween-20 (0.05%) for 1 minute per wash. Examine under a microscope fitted with a FITC filter.

Optimal working dilutions must be determined by end user.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Neural Stem Cells
This Anti-Stage-Specific Embryonic Antigen-1 Antibody, clone MC-480, Alexa Fluor 488 is validated for use in FC, IC, IF for the detection of Stage-Specific Embryonic Antigen-1.

Physical form

Purified immunoglobulin conjugated to Alexa Fluor 488. Liquid in Phosphate buffer with 15 mg/mL BSA as a stabilizer and 0.1% sodium azide

Storage and Stability

Maintain refrigerated at 2–8°C protected from light in undiluted aliquots for up to 24 months.

Analysis Note

Control
P19 mouse embryonic carcinoma cells

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effects of mechanical loading on tendons--an in vivo and in vitro model study.
Zhang, J; Wang, JH
Testing null
Z Cao et al.
The Journal of biological chemistry, 276(24), 21714-21723 (2001-03-30)
In this study we demonstrate that in corneal epithelium there is cell-cell contact-regulated expression of a 145-kDa glycoprotein (GP) bearing the glycan determinant Lewis(x) (Le(x)) (Galbeta(1,4)[Fucalpha(1,3)]GlcNAc). This glycoprotein (Le(x)-GP) was expressed in confluent/contact-inhibited cultures but not in sparse cultures of
M Boudjelal et al.
Genes & development, 11(16), 2052-2065 (1997-08-15)
We report the cDNA cloning of Stra13, a novel retinoic acid (RA)-inducible gene from P19 embryonal carcinoma cells that encodes a basic helix-loop-helix (bHLH) protein that shows the highest sequence similarities to the Drosophila Hairy and Enhancer of split and
Seema C Parte et al.
Stem cells and development, 26(24), 1781-1803 (2017-10-29)
Ovarian cancer is most lethal among gynecological cancers with often fatal consequences due to lack of effective biomarkers and relapse, which propels ovarian cancer research into unique directions to establish solid targeted therapeutics. "Ovarian stem cells" expressing germline pluripotent markers
S Vassilieva et al.
Experimental cell research, 258(2), 361-373 (2000-07-18)
Here, we demonstrate long-term cultivation of alkaline phosphatase-positive rat embryonic stem-like (RES) cell lines. RES cells were characterized by their typical growth in highly compacted cell clusters, which were found to be sensitive against enzymatic dissociation. RES cells expressed stage-specific

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service