HG000C68
Yeast Extract Agar
pH 7.2±0.2 (25 °C, after autoclaving)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352111
Recommended Products
composition
Agar, 15.0 g/L
Peptone, 5.0 g/L
Yeast Extract, 3.0 g/L
packaging
poly bottle of 12 × 500 g (HG000C68.500)
pH
7.2±0.2 (25 °C, after autoclaving)
storage temp.
15-25°C
Application
This medium is recommended for the plate count of micro-organisms in water.
Preparation Note
Suspend 23g in 1 litre demineralised water. Boil whilst stirring until completely dissolved. Autoclave at 121°C for 15 minutes.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service