Skip to Content
Merck
All Photos(2)

Key Documents

91823

Supelco

Gemfibrozil

analytical standard

Synonym(s):

2,2-Dimethyl-5-(2,5-dimethylphenoxy)pentanoic acid, 2,2-Dimethyl-5-(2,5-xylyloxy)valeric acid, 5-(2,5-Dimethylphenoxy)-2,2-dimethylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O3
CAS Number:
Molecular Weight:
250.33
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1ccc(C)c(OCCCC(C)(C)C(O)=O)c1

InChI

1S/C15H22O3/c1-11-6-7-12(2)13(10-11)18-9-5-8-15(3,4)14(16)17/h6-7,10H,5,8-9H2,1-4H3,(H,16,17)

InChI key

HEMJJKBWTPKOJG-UHFFFAOYSA-N

Gene Information

human ... PPARA(5465)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

C M Spencer et al.
Drugs, 51(6), 982-1018 (1996-06-01)
Gemfibrozil improves lipid and apolipoprotein profiles, particularly very low density lipoprotein (VLDL) triglyceride and high density lipoprotein (HDL) cholesterol levels, in patients with dyslipidaemia when administered at a total daily dose of 900 or 1200 mg. As demonstrated by the
Avik Roy et al.
Immunopharmacology and immunotoxicology, 31(3), 339-351 (2009-08-22)
Gemfibrozil is long known for its ability to reduce the level of triglycerides in the blood circulation and to decrease the risk of hyperlipidemia. However, a number of recent studies reveal that apart from its lipid-lowering effects, gemfibrozil can also
G J Magarian et al.
Archives of internal medicine, 151(9), 1873-1874 (1991-09-01)
Drug-induced myopathy has been reported with use of lovastatin, nicotinic acid, and clofibrate. A particularly severe form, often accompanied by rhabdomyolysis, has been reported with the use of lovastatin and gemfibrozil; however, as far as we know, no case has
P Zimetbaum et al.
Journal of clinical pharmacology, 31(1), 25-37 (1991-01-01)
Fibric acid derivatives (FADs) are a class of drugs that have been shown to reduce the production of very low-density lipoprotein (VLDL) while enhancing VLDL clearance due to the stimulation of lipoprotein lipase activity. The drugs can reduce plasma triglyceride
P A Todd et al.
Drugs, 36(3), 314-339 (1988-09-01)
Gemfibrozil is a lipid-regulating agent which is generically classified as a fibric acid derivative, but which exhibits different pharmacological effects from other such drugs. Published data indicate that in patients with all types of dyslipidaemia (except type I) gemfibrozil 800

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service