Skip to Content
Merck
All Photos(1)

Key Documents

41090

Supelco

2,4-Dimethylpentane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(CH3)2
CAS Number:
Molecular Weight:
100.20
Beilstein:
1696855
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.48 (vs air)

vapor pressure

3.29 psi ( 37.7 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

~7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.381 (lit.)
n20/D 1.381

bp

80 °C (lit.)

mp

−123 °C (lit.)

density

0.673 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C)CC(C)C

InChI

1S/C7H16/c1-6(2)5-7(3)4/h6-7H,5H2,1-4H3

InChI key

BZHMBWZPUJHVEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jaiho Cho et al.
Chemosphere, 68(2), 266-273 (2007-02-16)
The biodegradability of alkylate compounds in serum bottles was investigated in the presence and absence of ethanol or benzene, toluene, ethylbenzene, and p-xylene (BTEX). The biomass was acclimated to three different alkylates, 2,3-dimethylpentane, 2,4-dimethylpentane and 2,2,4-trimethylpentane in porous pot reactors.
M B Snipes et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 16(1), 81-91 (1991-01-01)
This paper describes apparatus and methods for measuring uptake of inhaled vapors in the nose and lungs of dogs. The system allows sampling of air from the trachea at specific times during inspiration and expiration without surgical manipulation of the
A R Dahl et al.
Toxicology and applied pharmacology, 109(2), 263-275 (1991-06-15)
The site of uptake of inhaled vapors profoundly influences which respiratory tract tissues receive the highest doses. How the site of uptake depends on the physicochemical properties of inhaled vapors has been the subject of experiment and speculation for decades

Protocols

This application demonstrates the suitability of the Petrocol column for the efficient analysis of hydrocarbons as listed in ASTM method D5134.

Protocol for GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH

-Xylene; Nonane; Propylbenzene; Mesitylene; 1,2,4-Trimethylbenzene; 1,2,3-Trimethylbenzene; 1,3-Diethylbenzene; 1,4-Dimethyl-2-ethylbenzene; 1,2-Dimethyl-4-ethylbenzene; Durene; 1,2,3,5-Tetramethylbenzene; 1,2,3,5-Tetramethylbenzene; 2-Methylnaphthalene (β)

GC Analysis of Hydrocarbons in Gasoline on Petrocol® DH, Isothermal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service