GC Analysis of Hydrocarbons in Gasoline on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
CONDITIONS
column
Equity-1, 15 m x 0.10 mm I.D., 0.10 μm (28039-U)
oven
40 °C (1 min), 45 °C/min to 150 °C (2 min)
inj. temp.
175 °C
detector
FID, 175 °C
carrier gas
hydrogen, 45 cm/sec constant
injection
0.1 μL 300:1 split
liner
2 mm I.D., straight
sample
Unleaded gasoline (refinery standard), neat
Description
Informations légales
null