Skip to Content
Merck
All Photos(2)

Key Documents

32738

Supelco

Azaperol

VETRANAL®, analytical standard

Synonym(s):

α-(4-Fluorophenyl)-4-(2-pyridinyl)-1-piperazinebutanol, 1-(4-Fluorophenyl)-4-(4-pyridin-2-yl-piperazin-1-yl)butan-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24FN3O
CAS Number:
Molecular Weight:
329.41
Beilstein:
568396
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

OC(CCCN1CCN(CC1)c2ccccn2)c3ccc(F)cc3

InChI

1S/C19H24FN3O/c20-17-8-6-16(7-9-17)18(24)4-3-11-22-12-14-23(15-13-22)19-5-1-2-10-21-19/h1-2,5-10,18,24H,3-4,11-15H2

InChI key

LVXYAFNPMXCRJI-UHFFFAOYSA-N

General description

Azaperol is one of the major metabolites of azaperone which is used as a tranquilizer.

Application

Azaperol may be used as an analytical reference standard for the determination of the analyte in:
  • Pig tissues by liquid chromatography–tandem mass spectrometry (LC-MS/MS).
  • Egg, fish and meat samples by high-resolution liquid chromatography coupled to time-of-flight mass spectrometry (HRLC-TOF-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Fluchard et al.
Journal of chromatography. B, Biomedical sciences and applications, 744(1), 139-147 (2000-09-14)
A quick, simple method for quantifying carazolol, azaperol and azaperone is described. Liquid extraction was followed by a clean-up on an Oasis SPE cartridge. The analytes were separated by HPLC and analysed by MS-MS with atmospheric pressure chemical ionisation in
Multi-residue screening of veterinary drugs in egg, fish and meat using high-resolution liquid chromatography accurate mass time-of-flight mass spectrometry.
Peters RJB, et al.
Journal of Chromatography A, 1216(46), 8206-8216 (2009)
M L Olmos-Carmona et al.
Journal of chromatography. B, Biomedical sciences and applications, 734(1), 113-120 (1999-11-26)
A method for analysis of veterinary tranquillizers in urine using gas chromatography-mass spectrometry (GC-MS) is described. Detection limits are 5 microg/l for ketamine, azaperone and the phenothiazines (chlor-, aceto- and propionylpromazine), 10 microg/l for haloperidol, 20 microg/l for xylazine and
J C Jaen et al.
Journal of medicinal chemistry, 36(24), 3929-3936 (1993-11-26)
The enantiomers of reduced haloperidol (3a), azaperol (3b), and the related compound BMY-14802 (3c) were prepared in high optical purity. The affinity of these compounds for dopamine D2 and D3 receptors, and sigma S1 and S2 sites was determined in
W Arneth
Zeitschrift fur Lebensmittel-Untersuchung und -Forschung, 201(3), 261-265 (1995-09-01)
A method is described for the analytical determination of the neuroleptics azaperone (plus its metabolite azaperol), acetylpromazine, propionylpromazine, chlorpromazine and the beta-blocking agent carazolol in pork kidneys. These compounds may be used illegally to calm pigs during their transport to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service