Documentos clave
EP0030132548
Eppendorf® Twin tec real-time PCR plates
white plate, conical bottom white wells, semi-skirted, pkg of 25 ea
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
conical bottom white wells
white plate
size:
96 well
feature:
skirt (Semi skirted)
material
conical bottom white wells
white plate
feature
skirt (Semi skirted)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030132548
technique(s)
PCR: suitable
size
96 well
working volume
250 μL
General description
The limiting factor in low-volume real-time PCR (qPCR) is often the remaining fluorescence intensity. Eppendorf twin.tec real-time PCR plates are specially manufactured for qPCR, as they offer all the advantages of regular twin.tec plates, but with the added advantage of white wells. These reflect fluorescence much better than clear or frosted wells, so lower levels of fluorescence are still detectable with the same instrument – just by using the right consumables. The qPCR plates′ white wells also significantly reduce interfering background fluorescence and lead to increased consistency between replicates, and giving highly reproducible results. The wells are made from polypropylene – which guarantees fast heat transfer for fast assays – and the frame is made of rigid polycarbonate, to ensure easy and reliable handling – both in manual and automated assays.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)