553215
RAPIDstain Reagent
An ultrasensitive Coomassie-based reagent for staining polyacrylamide gels.
About This Item
UNSPSC Code:
12171500
Productos recomendados
Quality Level
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
assay range
sensitivity: 4-8 ng
(BSA)
shipped in
ambient
storage temp.
15-25°C
General description
An ultrasensitive Coomassie-based reagent for staining polyacrylamide gels. No mixing or preparation is involved, no fixation step is needed, and no destaining is required. RAPIDstain Reagent stains only protein, within 5-10 min, and leaves a crystal-clear background with high resolution. Contain the following component:
• RAPIDstain 1 x 1 liter Suitable for 20 mini gels (8 x 10 cm) or 10 larger gels (12 x 15 cm)
• RAPIDstain 1 x 1 liter Suitable for 20 mini gels (8 x 10 cm) or 10 larger gels (12 x 15 cm)
Warning
Toxicity: Harmful (C)
Other Notes
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.