MilliporeSigma
357006
Sigma-Aldrich
Tantalum
wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sinónimos:
Ta
About This Item
Fórmula empírica (notación de Hill):
Ta
Número de CAS:
Peso molecular:
180.95
EC Number:
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
<0.01 mmHg ( 537.2 °C)
Quality Level
assay
≥99.9% trace metals basis
form
wire
autoignition temp.
572 °F
resistivity
13.5 μΩ-cm, 20°C
diam.
0.5 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
Quantity
1 m (approximately 3.3 g)
Storage Class
11 - Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Orbital ultrasound in the selective screening of a dural-cavernous sinus fistula.
Hamid Shokoohi et al.
European journal of emergency medicine : official journal of the European Society for Emergency Medicine, 20(2), 137-139 (2013-02-26)
S Nag et al.
Journal of the mechanical behavior of biomedical materials, 16, 21-28 (2012-11-10)
Microstructure and mechanical properties of laser deposited complex quaternary Ti-34Nb-7Zr-7Ta (all wt%), an orthopedic load-bearing implant alloy, has been investigated in detail in both as-deposited as well as heat-treated (β-solutionized and quenched) conditions. The difference in stress-strain behavior of this
Thomas Larzon et al.
Vascular, 21(2), 87-91 (2013-03-20)
Control of back bleeding from the hypogastric artery into the aneurysm after endovascular aneurysm repair (EVAR) of a ruptured aorto-iliac aneurysm may be necessary in order to avoid a type II endoleak. It is an emergency situation and selective catheterization
Stuart A Callary et al.
Clinical orthopaedics and related research, 471(7), 2238-2244 (2013-01-22)
One cross-linked polyethylene (XLPE) liner is manufactured using a lower dose of radiation, 5 Mrad, which may result in less cross-linking. The reported in vivo wear rate of this XLPE liner in patients undergoing THA has varied, and has included
Shi-Hua Yu et al.
Water environment research : a research publication of the Water Environment Federation, 84(12), 2115-2122 (2013-01-25)
Tantalum hydroxide exhibits the ability for the removal of phosphate from aqueous solution. The kinetic study, adsorption isotherm, thermodynamic study, desorption, and foreign anions effect were examined in batch experiments. The kinetic process was very well described by a pseudo-second-order
Artículos
Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.