重要文件
Z568961
Sigma-Aldrich
Aldrich® Liebig condenser with cup
Overall H 200 mm, joint: ST/NS 14/20
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
接合處
joint: ST/NS14/20
總高度
200 mm
尋找類似的產品? 前往 產品比較指南
一般說明
Condensate cup catches water that forms on the outer surface of the condenser during use and eliminates puddling on reactor and equipment. Tubing connects to cup′s hose barb to drain away water automatically.
法律資訊
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
從最近期的版本中選擇一個:
分析證明 (COA)