重要文件
PL800GANEO-10
Plas-Labs replacement neoprene gloves
for 2 piece glove set, size 10 (for 8 in. ports)
登入查看組織和合約定價
About This Item
分類程式碼代碼:
46181504
NACRES:
NB.35
材料
neoprene
尺寸
10 (for 8 in. ports)
特點
reusable
for use with glove box sleeves
包裝
pack of 1 pair
製造商/商標名
Plas-Labs 800GANEO-10
尋找類似的產品? 前往 產品比較指南
一般說明
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
  • White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
  • Black neoprene rubber for chemical resistance
  • Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
  • Green nitrile rubber for resistance to oil, fuel, and acids
  • Latex rubber for flexibility
法律資訊
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
從最近期的版本中選擇一個:
分析證明 (COA)