重要文件
MT51340292
Mettler Toledo InLab® 605 dissolved oxygen sensor
cable L 10 m
登入查看組織和合約定價
About This Item
分類程式碼代碼:
41115600
NACRES:
NB.79
材料
glass fiber shaft (reinforced)
membrane (silicone/PTFE)
描述
BNC/RCA (Cinch), IP67 connector
NTC22kΩ temperature probe
interchangeable membrane bodies
製造商/商標名
Mettler-Toledo 51340292
電纜長度
10 m
尋找類似的產品? 前往 產品比較指南
一般說明
The InLab® 605 DO sensor incorporates the newest technology in polarographic dissolved oxygen determination. With the InLab® 605 dissolved oxygen sensor you will find the high accuracy and reproducibility that you have come to trust with METTLER TOLEDO products.
The InLab® 605 sensor can be used in combination with the SevenExcellence multichannel benchtop meter or the SevenGo 6 portable meters.
There are certain applications in which such sensors need to be rugged enough to withstand harsh conditions in factories, laboratories or out in the field. For this reason the membrane is protected by a steel mesh and the shaft is made out of glass fiber reinforced PPS (polyphenylene sulfide).
To go with the InLab® 605 METTLER TOLEDO offers the best of the best accessories: membrane kit, electrolyte, sealing kit and electrode weight.
法律資訊
InLab is a registered trademark of Mettler-Toledo, Inc.
從最近期的版本中選擇一個:
分析證明 (COA)