527793
2,3-Dichlorobenzonitrile
Synonym(s):
2,3-Dichlorophenyl cyanide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Cl2C6H3CN
CAS Number:
Molecular Weight:
172.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
solid
mp
60-64 °C (lit.)
SMILES string
Clc1cccc(C#N)c1Cl
InChI
1S/C7H3Cl2N/c8-6-3-1-2-5(4-10)7(6)9/h1-3H
InChI key
OHDYZVVLNPXKDX-UHFFFAOYSA-N
General description
2,3-Dichlorobenzonitrile is formed as an intermediate during the synthesis of 2,3-dichlorobenzoic acid.
Application
2,3-Dichlorobenzonitrile may be used to synthesize 2-fluoro-3-chlorobenzonitrile and 6-amino-10-chlorobenzo[c]quinolizinium chloride.
Reactant involved in:
- Nucleophilic aromatic fluorination
- Reactions with magnesium amides for synthesis of carboxamides
- Suzuki-Miyaura coupling reactions
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Room-temperature nucleophilic aromatic fluorination: experimental and theoretical studies.
Haoran Sun et al.
Angewandte Chemie (International ed. in English), 45(17), 2720-2725 (2006-03-21)
Preparation of 2, 3-Dichlorobenzoic acid.
Deng H, et al.
Chinese Journal of Pharmaceuticals / Chung-kuo i yao kung yeh tsa chih, 3, 006-006 (2005)
F Becq et al.
The Journal of biological chemistry, 274(39), 27415-27425 (1999-09-17)
Chloride channels play an important role in the physiology and pathophysiology of epithelia, but their pharmacology is still poorly developed. We have chemically synthesized a series of substituted benzo[c]quinolizinium (MPB) compounds. Among them, 6-hydroxy-7-chlorobenzo[c]quinolizinium (MPB-27) and 6-hydroxy-10-chlorobenzo[c]quinolizinium (MPB-07), which we
Jishi Wang et al.
Journal of chromatography. A, 1521, 10-18 (2017-09-25)
In this research, the manual two-step QuEChERS approach has been streamlined and automated into a one-step method using a cleanup tube fitted within an extraction tube. A novel automatic QuEChERS combination have been developed to simplify the QuEChERS procedures and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service