Merck
20437-U
Supelco
塑料色谱柱帽
for use with 1/8 in. O.D. columns
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
SB.54
包裝
pkg of 100 ea
接頭
female cap for 1/8 in. O.D. column (slip-on)
相容性
for use with 1/8 in. O.D. columns
正在寻找类似产品? 访问 产品对比指南
一般說明
These plastic caps will easily slip over the ends of packed GC columns to protect them during storage.
應用
Plastic column cup maybe used to close the immunoaffinity chromatography column used for separating cross-linked protein complexes adhering to P1 protein of Mycoplasma pneumonia.
历史批次信息供参考:
分析证书(COA)
Lot/Batch Number
抱歉,我们目前尚未线上提供该产品的COA。
访问文档库
G Layh-Schmitt et al.
Microbiology (Reading, England), 146 ( Pt 3), 741-747 (2000-04-04)
Adherence of Mycoplasma pneumoniae to host cells requires several mycoplasmal membrane proteins and cytoskeleton-like proteins in addition to the adhesin P1, a transmembrane protein of 170 kDa. To analyse interactions of the P1 adhesin with other membrane proteins or with