主要文件
Z692611
microDOC Compact gel documentation system
AC/DC input 230 V AC
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41105300
NACRES:
NB.22
特點
Contains UK and Schuko plugs
製造商/商標名
Cleaver CSI-MICRODOC
正在寻找类似产品? 访问 产品对比指南
一般說明
The microDOC Compact Gel Documentation System gives you all of the advantages of the Basic system with the advantage of a large 8 inch TFT colour monitor allowing clear and simple image visualization. Can be used with most 12 x 12 cm tranilluminators.
Available with optional TotalLab 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
法律資訊
TotalLab is a trademark of Nonlinear Dynamics Ltd.
历史批次信息供参考:
分析证书(COA)