主要文件
WHA2200-654
Whatman® 1PS phase separator filter papers
W × L (40 mm) (50 m), pkg of 5 pkg
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41115727
NACRES:
NB.24
包裝
pkg of 5 pkg
寬度 × 長度
(40 mm) (50 m)
正在寻找类似产品? 访问 产品对比指南
一般說明
Whatman 1PS Phase Separator from Cytiva′s business is a hydrophobic filter for routine solvent extraction. Within seconds it retains aqueous phases, while solvent phases flow through.
Hydrophobic solvent extraction paper
Solvent extraction can be time-consuming and messy when numerous separations are required. Cytiva′s Whatman 1PS Phase Separator is impregnated with stabilized silicone, creating a hydrophobic filter paper that can replace separatory funnel use in certain applications.
Rapid solvent extraction
Mixed phases are poured directly into the Whatman 1PS Phase Separator filter. The solvent phase flows through the paper quickly and cleanly, automatically stopping after the process is complete; all of the aqueous phase is retained. Droplets are automatically separated within seconds.
特點和優勢
  • Impregnated with stabilized silicone to create a hydrophobic filter
  • Multiple separations can be processed in parallel
  • No special training required
  • Can replace a separatory funnel in certain applications
法律資訊
Whatman is a registered trademark of Cytiva
历史批次信息供参考:
分析证书(COA)