Skip to Content
Merck
All Photos(1)

Key Documents

G004997

HPLC Analysis of Homatropine Enantiomers on Astec® Cellulose DMP

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase: [A] methanol; [B] DEA; (100:0.1, A:B)
flow rate: 0.5 mL/min
column temp.: 25 °C
detector: UV, 230 nm
injection: 2 μL
sample: 2 mg/mL in mobile phase

suitability

application for HPLC

application(s)

pharmaceutical (small molecule)

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    DL-Homatropine

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service