Skip to Content
Merck
All Photos(2)

Key Documents

MAK221

Sigma-Aldrich

Beta-Lactamase Activity Assay Kit

Sufficient for 100 Colorimetric tests

Synonym(s):

β-Lactamase Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

detection method

colorimetric

storage temp.

−20°C

General description

β-Lactamase (bL; EC 3.5.2.6) is an enzyme first identified in Escherichia coli and has been described as penicillinase. A number of βLs have since been identified from various bacteria. βLs specifically hydrolyze β-lactam rings present in antibiotics such as penicillin, cephalosporins, monobactam, and carbapenem, and confer resistance against these antibiotics.

Suitability

Suitable for the detection of β-lactamase activity in biological samples, food, and fermentation samples.

Principle

The β-Lactamase Activity Assay Kit provides a simple assay for measuring βL activity in βL-secreting bacteria in fermentation media and bacterial cultures. It can also be used to detect βL-secreting bacteria in saliva, urine, and serum of infected mammals and in food samples. βL activity is measured by hydrolyzing a chromogenic cephalosporin called nitrocefin, producing a colorimetric product (A490), proportional to the enzymatic activity present. One unit of β-lactamase is the amount of enzyme required to hydrolyze 1.0 μmole of nitrocefin per minute at pH 7.0 at 25 °C.

Kit Components Only

Product No.
Description

  • β-Lactamase Assay Buffer

  • Nitrocefin, in DMSO

  • Positive Control

  • βL Hydrolysis Buffer

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Class C β-lactamases operate at the diffusion limit for turnover of their preferred cephalosporin substrates.
Bulychev A and Shahriar M
Antimicrobial Agents and Chemotherapy, 43(7), 1743-1746 (1999)
Jennifer M Colquhoun et al.
mBio, 12(6), e0313721-e0313721 (2021-12-08)
β-Lactamase expression is the major mechanism of resistance to penicillins, cephalosporins, and carbapenems in the multidrug-resistant (MDR) bacterium Acinetobacter baumannii. In fact, stable high-level expression of at least one β-lactamase has been rapidly increasing and reported to occur in up
Beta-lactam antibiotics: from antibiosis to resistance and bacteriology.
Kong K F, et al.
Acta Pathologica, Microbiologica et Immunological Scandinavica, 118(1), 1-36 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service