NIST640F
Line Position And Line Shape Standard for Powder Diffraction
(Silicon powder), NIST® SRM® 640F
Synonym(s):
Silicon powder, Silicone powder
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116107
Recommended Products
form
powder
packaging
pkg of 7.5 g
manufacturer/tradename
NIST®
technique(s)
diffraction/scattering: suitable
General description
The Standard Reference Material (SRM) was prepared from ultra-high purity, intrinsic silicon boules that were crushed and jet milled to a median particle size of 4.1μm. The resulting powder was then annealed under gettered argon at 1000 °C for two hours and bottled under argon.
SRM 640F_cert
SRM 640F_SDS
SRM 640F_cert
SRM 640F_SDS
Application
This SRM is intended for use as a standard for calibration of diffraction line positions and line shapes, determined through powder diffractometry.
Features and Benefits
- A unit of SRM 640f consists of approximately 7.5 g of silicon powder bottled under argon.
- Analysis of X-ray powder diffraction data indicated that the SRM material is homogeneous with respect to diffraction properties.
- SRM 640f was bottled under argon to protect against humidity.
- The certification data was analyzed using the FPA method with Pawley refinements as implemented in TOPAS.
- Expiration details and storage instructions are available in the NIST certificate.
Other Notes
Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 2
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service