Skip to Content
Merck
All Photos(1)

Key Documents

575305-M

Millipore

T0070907

A cell-permeable chloro-nitro-benzamido compound that acts as a potent, specific, irreversible, and high-affinity antagonist of PPARγ with a Ki of 1 nM.

Synonym(s):

T0070907, 2-Chloro-5-nitro-N-(4-pyridyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8ClN3O3
CAS Number:
Molecular Weight:
277.66
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

InChI

1S/C12H8ClN3O3/c13-11-2-1-9(16(18)19)7-10(11)12(17)15-8-3-5-14-6-4-8/h1-7H,(H,14,15,17)

InChI key

FRPJSHKMZHWJBE-UHFFFAOYSA-N

General description

A cell-permeable chloro-nitro-benzamido compound that acts as a potent, specific, irreversible, and high-affinity antagonist of PPARγ with a Ki of 1 nM. Displays >800-fold greater selectivity for PPARγ over PPARα and PPARδ (Ki = 0.85 µM and 1.8 µM, respectively). Blocks hormone- and agonist-induced adipogenesis in 3T3-L1 cells. It suppresses interactions between PPARγ and coactivator-derived peptides, while promotes the recruitment of corepressor-derived peptides. Shown to modulate the interaction of PPARγ2 with the cofactor proteins through covalent binding to Cys313 in its ligand-binding domain.
A cell-permeable, potent, specific, irreversible, and high-affinity antagonist of PPARγ (Ki = 1 nM). Reported to be >800-fold more specific for PPARγ compared to PPARα (Ki = 850 nM) and PPARδ (Ki = 1.8 µM). Shown to block hormone- and agonist-induced adipogenesis in 3T3-L1 cells. Suppresses interactions between PPARγ and coactivator-derived peptides while promoting the recruitment of co-repressor-derived peptides. Also shown to modulate the interaction of PPARγ2 with cofactor proteins through covalent binding to Cys313 in the ligand-binding domain.

Biochem/physiol Actions

Primary Target
PPARγ
Target Ki: 1 nM for PPARγ

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lee, G., et al. 2002. J. Biol. Chem.277, 19649.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service